Learn More
Hach Company Inductive Conduc. sensor PP
Catalog No. 5000207782 Shop All Hach Company Products
Encompass
Digital Sanitary Inductive (Electrodeless) Conductivity Sensor with PP Body MaterialPolypropylene Sanitary (CIP) Flange Mounting Style 6m (20 ft) Analog Cable Electrodeless Conductivity Sensor includes 6120800 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3 ft) Digital Extension Cable
Catalog No. 50-002-07782 Supplier Hach Company Supplier No. D3705E2T
Digital Sanitary Inductive (Electrodeless) Conductivity Sensor with PP Body MaterialPolypropylene Sanitary (CIP) Flange Mounting Style 6m (20 ft) Analog Cable Electrodeless Conductivity Sensor includes 6120800 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3 ft) Digital Extension Cable