Learn More
Hach Company Inductive Conduc. sensor PVDF
Catalog No. 5000207720 Shop All Hach Company Products
Encompass
Digital Sanitary Inductive-Electrodeless Conductivity Sensor for Low pH applications-PVDF Body MaterialPVDF Sanitary (CIP) Flange Mounting Style 6m (20 ft) Analog Cable Electrodeless Conductivity Sensor includes 6120800 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3 ft) Digital Extension Cable
Catalog No. 50-002-07720 Supplier Hach Company Supplier No. D3706E2T
Digital Sanitary Inductive-Electrodeless Conductivity Sensor for Low pH applications-PVDF Body MaterialPVDF Sanitary (CIP) Flange Mounting Style 6m (20 ft) Analog Cable Electrodeless Conductivity Sensor includes 6120800 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3 ft) Digital Extension Cable