Learn More
Hach Company Inductive sensor-convertible
Catalog No. 5000207596 Shop All Hach Company Products
Encompass
Digital Inductive Conductivity Sensor-Convertible Body StylePolypropylene Convertible Mounting Style 6m (20ft) Analog Cable Electrodeless Conductivity Sensor includes 6120800 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3ft) Digital Extension Cable
Catalog No. 50-002-07596 Supplier Hach Company Supplier No. D3725E2T
Digital Inductive Conductivity Sensor-Convertible Body StylePolypropylene Convertible Mounting Style 6m (20ft) Analog Cable Electrodeless Conductivity Sensor includes 6120800 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3ft) Digital Extension Cable