Learn More
New England Biolabs, Inc. Induro Reverse Transcriptase - 10000 units
SDP
Catalog No. 502269745
Encompass
Induro Reverse Transcriptase is a group II intron-encoded RT that exhibits increased thermostability high processivity and increased tolerance of inhibitors in the synthesis of cDNA from RNA. It is an ideal enzyme for challenging cDNA synthesis reactions from long transcripts RNAs with strong secondary structures and RNA samples with inhibitors. Additionally Induro Reverse Transcriptase supports RNA-seq applications such as long-read RNA sequencing.
This item is not returnable. View return policy
Catalog No. 50-226-9745 Supplier New England Biolabs, Inc. Supplier No. M0681L
Induro Reverse Transcriptase is a group II intron-encoded RT that exhibits increased thermostability high processivity and increased tolerance of inhibitors in the synthesis of cDNA from RNA. It is an ideal enzyme for challenging cDNA synthesis reactions from long transcripts RNAs with strong secondary structures and RNA samples with inhibitors. Additionally Induro Reverse Transcriptase supports RNA-seq applications such as long-read RNA sequencing.