Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI Industrial Amber Evaporation Flask
SDP

Catalog No. 05405228 Shop All BUCHI Products
Change view
Click to view available options
Volume (Metric):
10 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
05-405-228 10 L
05-405-229 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-405-228 Supplier BUCHI Supplier No. 11069604
Add to Cart
Add to Cart

Amber evaporating flasks for use with industrial rotary evaporators when UV-protection is important.

  • Amber colored glass provides UV-protection
  • For use with rotary evaporators
  • 150 mm Flange

Specifications

Color Amber
Material Borosilicate Glass (3.3)
Type Evaporating Flask
For Use With (Application) Evaporating Light-Sensitive Samples
For Use With (Equipment) R-220 Pro, R-220 Essential, and R-220 Rotary Evaporators
Volume (Metric) 10 L
Diameter (Metric) 150 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.