Learn More
MSC Atrix HEPA Vacuum, 1000 Hour Motor, 1 Gallon Capacity, 3 Levels of Filtration
Catalog No. 502838321
Encompass
The Atrix Omega Green Supreme IPM HEPA Vacuum is specifically configured for Pest Management Professionals. This robust vacuum features a durable 1,000-hour rated Ametek Advantek II motor, a 1 gallon capacity, and three levels of filtration, ensuring thorough cleaning and containment of pests. Its design includes self-storage for accessories, making it convenient for professional use.
  • 1,000 hour rated Ametek Advantek II motor
  • 25 ft detachable power cord
  • 1 gallon capacity
  • Features 3 levels of filtration
  • Self-storage and contained accessories
  • Maximum amperage of 5.2 A
  • Includes extension wands, crevice tool with brush, 16 in flex wand, shoulder strap, 6 ft clear hose, HEPA filter, filter and hose plugs, and utility nozzle
Catalog No. 50-283-8321 Supplier MSC Supplier No. 71272710
The Atrix Omega Green Supreme IPM HEPA Vacuum is specifically configured for Pest Management Professionals. This robust vacuum features a durable 1,000-hour rated Ametek Advantek II motor, a 1 gallon capacity, and three levels of filtration, ensuring thorough cleaning and containment of pests. Its design includes self-storage for accessories, making it convenient for professional use.
  • 1,000 hour rated Ametek Advantek II motor
  • 25 ft detachable power cord
  • 1 gallon capacity
  • Features 3 levels of filtration
  • Self-storage and contained accessories
  • Maximum amperage of 5.2 A
  • Includes extension wands, crevice tool with brush, 16 in flex wand, shoulder strap, 6 ft clear hose, HEPA filter, filter and hose plugs, and utility nozzle