Learn More
BURKLE INC Industrial bucket, LDPE white, withmetal handle, 4.49 gal. (17l)
Catalog No. 5000025947
Encompass_Preferred
Industrial bucket for transporting, decanting and mixing. Safe emptying with wide spout. Handle made of metal.
Catalog No. 50-000-25947 Supplier BURKLE INC Supplier No. 23081700
Industrial bucket for transporting, decanting and mixing. Safe emptying with wide spout. Handle made of metal.