Learn More
MSC Safety Speed Industrial Vacuum, 26 Gallon, 3 HP, 3-Stage Filtration
Catalog No. 502838472
Encompass
This industrial vacuum offers high-performance dust collection with a dual-headed dust port for connecting to two machines, capturing over 95% of dust particles. It features up to 3 stages of filtration, including an optional H14 HEPA filter that exceeds OSHA's respiratory criteria. Each unit is individually certified through extensive quality control.
  • 26 gallon collection capacity with a 3 HP motor.
  • Up to 3 filtration stages, including a 3-micron Class L main filter and optional 0.18 micron H14 HEPA filter.
  • H14 HEPA filter provides 99.995% collection efficiency and exceeds OSHA respiratory criteria.
  • Dual-headed dust port for versatile machine connection.
  • Individually certified through a rigorous quality control process.
  • Maximum water lift of 80.9 inH2O and airflow up to 282.5 CFM.
  • 1.5 in diameter dust collection port.
Catalog No. 50-283-8472 Supplier MSC Supplier No. 67493783
This industrial vacuum offers high-performance dust collection with a dual-headed dust port for connecting to two machines, capturing over 95% of dust particles. It features up to 3 stages of filtration, including an optional H14 HEPA filter that exceeds OSHA's respiratory criteria. Each unit is individually certified through extensive quality control.
  • 26 gallon collection capacity with a 3 HP motor.
  • Up to 3 filtration stages, including a 3-micron Class L main filter and optional 0.18 micron H14 HEPA filter.
  • H14 HEPA filter provides 99.995% collection efficiency and exceeds OSHA respiratory criteria.
  • Dual-headed dust port for versatile machine connection.
  • Individually certified through a rigorous quality control process.
  • Maximum water lift of 80.9 inH2O and airflow up to 282.5 CFM.
  • 1.5 in diameter dust collection port.