Learn More
MSC Safety Speed Automated Vacuum, 13 Gallon, 2 HP, 115/220 V
Catalog No. 502838474
Encompass
This 13-gallon automated dry vacuum is designed for commercial use, featuring a single-phase, two-horsepower, two-stage bypass motor. Operating on 115 or 220 volts, it includes an automated on/off switch that syncs with connected machinery, efficiently collecting material down to 3 microns.
  • 13 gallon collection capacity
  • 2 horsepower motor, 115/220 V, single phase
  • 110 CFM maximum airflow and 105 inch maximum water lift
  • Collects material down to 3 microns
  • Automated on/off function for seamless operation
  • Designed for high-quality, long-life commercial use
  • Uses universal filters
  • Includes 20 foot crushproof hose
Catalog No. 50-283-8474 Supplier MSC Supplier No. 67493791
This 13-gallon automated dry vacuum is designed for commercial use, featuring a single-phase, two-horsepower, two-stage bypass motor. Operating on 115 or 220 volts, it includes an automated on/off switch that syncs with connected machinery, efficiently collecting material down to 3 microns.
  • 13 gallon collection capacity
  • 2 horsepower motor, 115/220 V, single phase
  • 110 CFM maximum airflow and 105 inch maximum water lift
  • Collects material down to 3 microns
  • Automated on/off function for seamless operation
  • Designed for high-quality, long-life commercial use
  • Uses universal filters
  • Includes 20 foot crushproof hose