Learn More
Enterprise Technology Solutions Industrial Cleaner and Degreaser
SDP
Supplier:  Enterprise Technology Solutions SMP13005CT
Encompass_Preferred
Readily biodegradable formula.Trusted non-flammable, non-abrasive and non-corrosive cleaner is a safer alternative to caustic cleaners and solvents.Versatile formula can be used to clean and degrease all washable surfaces.Cost-effective concentrate can be used manually or with cleaning equipment.VOC compliant and exempt from OSHA and EPA special handling and personal protection requirements for hazardous materials.Butyl and phosphate-free formula.Ideal for a wide range of industries including manufacturing, MRO, food processing, mining, automotive, transportation and engineering.Cuts through tough grease, oils, cutting fluids, built-up grime and residue on equipment, engines, machinery, vehicles, manufactured parts and more.Cleans and deodorizes food residue, oils and grease from countertops, appliances, grills, vents, traps and trash cans.Perfect for surface prep prior to painting, plating or welding.
Catalog No. 50-211-1024