Learn More
MSC CRC Industrial Degreaser, Aerosol, Solvent-Based, Non-Chlorinated, 14 oz
Catalog No. 502869242
Encompass
Mech Force Industrial Degreaser is an aerosol blend of organic solvents and non-chlorinated components. It is designed to remove oil from various mechanical parts and equipment.
  • Aerosol form for easy application
  • Solvent-based formula for effective oil removal
  • Non-chlorinated blend of organic solvents
  • NSF K1 and K2 registered, Boeing D6-117487, and RoHS compliant
  • Suitable for air tools, bearings, chains, clutches, and more
  • High dielectric strength of 6600 V
Catalog No. 50-286-9242 Supplier MSC Supplier No. 53801726
Mech Force Industrial Degreaser is an aerosol blend of organic solvents and non-chlorinated components. It is designed to remove oil from various mechanical parts and equipment.
  • Aerosol form for easy application
  • Solvent-based formula for effective oil removal
  • Non-chlorinated blend of organic solvents
  • NSF K1 and K2 registered, Boeing D6-117487, and RoHS compliant
  • Suitable for air tools, bearings, chains, clutches, and more
  • High dielectric strength of 6600 V