Learn More
Cole-Parmer BrandTech Large industrial-size HDPE funnel, 12 L
Catalog No. NC9683140 Shop All Cole-Parmer Products
Encompass_Preferred
Low density polyethylene (LDPE) funnels feature external ribbing for air release and have a handle tab for easy lifting. Polypropylene and high density polypropylene (HDPE) funnels are translucent and show content levels. Polypropylene funnels are autoclavahle.
Catalog No. NC9683140 Supplier Cole-Parmer Supplier No. 0612100
Low density polyethylene (LDPE) funnels feature external ribbing for air release and have a handle tab for easy lifting. Polypropylene and high density polypropylene (HDPE) funnels are translucent and show content levels. Polypropylene funnels are autoclavahle.