Learn More
MSC Industrial Magnetics, Inc. Lift Magnet, 220 lb Capacity, Permanent
Catalog No. 5000530875
Encompass
This permanent magnet lift is designed for lifting round and curved surfaces such as shafts, thick-walled pipe, or billet, with the ability to also handle flat materials. It features specially designed pole shoes that provide a deep penetrating magnetic field and part stability, offering a 220 lb working load limit.
  • On/off rare earth permanent magnet
  • Handle requires only 90 degree rotation and locks at both on/off positions
  • Durable, recessed metal nameplates
  • Corrosion-resistant, plated magnet surface
  • Heat resistant up to 180°F (82°C)
  • 3:1 design factor
  • Meets ASME B30.20 BTH-1 Design Category B, Service Class 3
Catalog No. 50-005-30875 Supplier MSC Supplier No. 71760250
This permanent magnet lift is designed for lifting round and curved surfaces such as shafts, thick-walled pipe, or billet, with the ability to also handle flat materials. It features specially designed pole shoes that provide a deep penetrating magnetic field and part stability, offering a 220 lb working load limit.
  • On/off rare earth permanent magnet
  • Handle requires only 90 degree rotation and locks at both on/off positions
  • Durable, recessed metal nameplates
  • Corrosion-resistant, plated magnet surface
  • Heat resistant up to 180°F (82°C)
  • 3:1 design factor
  • Meets ASME B30.20 BTH-1 Design Category B, Service Class 3