Rectangular Parallel Pole Electromagnets offer on/off capability and can be controlled in an automated sheet metal handling system. They efficiently hold and release ferrous material on command through controlled DC electrical current. These electromagnets provide improved reach through air gaps and can accommodate parts with uneven surfaces or odd shapes using custom machined pole shoes. This model is a 24 V DC, 36 W electromagnet with a 430 lb lifting capacity and continuous duty, ideal for various industrial applications.
- Provides excellent magnetic holding power.
- Features 24 V DC operation with 36 W power consumption.
- Offers a maximum lifting capacity of 430 lb.
- Designed for continuous duty operation.
- Includes side-mounted terminal blocks for easy wiring.
- Equipped with two mounting holes with 5/16-18 thread.
- Compact dimensions for versatile integration.