Learn More
Cadence Inc Industrial Razor Blades, 1.5" X 0.315
SDP
Catalog No. NC1625245
Encompass
This item has a minimum order quantity of 10 per supplier requirements.
Industrial Razor Blades, 1.5" X 0.315" no hole, Endurium, zirconia ceramic, 0.012 in thickness
Catalog No. NC1625245 Supplier Cadence Inc Supplier No. CT927
This item has a minimum order quantity of 10 per supplier requirements.
Industrial Razor Blades, 1.5" X 0.315" no hole, Endurium, zirconia ceramic, 0.012 in thickness