Learn More
Mascon Medical Industrial Scanner / Handheld Barcode Scanner / Wired / iData / J16 / Multi-Spectral Scanning / Polarized Light Elimination / Fast Decoding Algorithm / USB Connection / Electronics And Industrial Manufacturing / Each
SDP
Catalog No. NC4031557
Encompass_Preferred
High-Performance Handheld Barcode Scanner Provides Accurate Data Capture In Electronics Manufacturing / Industrial Manufacturing / And Home Appliance Assembly. Features Multi-Spectral Scanning With Polarized Light Elimination And NPU-Accelerated Fast Decoding Capable Of Reading Low-Contrast And Difficult Barcodes Including DPM Codes. USB Plug & Play Connectivity Enables Quick Integration And High-Speed Barcode Capture. Reduces Reflection And Ghosting On Metal / Glass / And Ceramic Surfaces. Compact / Durable / IP52 Protection / Drop Resistance From 1.8m Ensures Reliable Operation In Industrial Environments. Ideal For Inventory Management / Electronics / Home Appliance / And Automotive Parts Handling.
Catalog No. NC4031557 Supplier Mascon Medical Supplier No. IMJ16
High-Performance Handheld Barcode Scanner Provides Accurate Data Capture In Electronics Manufacturing / Industrial Manufacturing / And Home Appliance Assembly. Features Multi-Spectral Scanning With Polarized Light Elimination And NPU-Accelerated Fast Decoding Capable Of Reading Low-Contrast And Difficult Barcodes Including DPM Codes. USB Plug & Play Connectivity Enables Quick Integration And High-Speed Barcode Capture. Reduces Reflection And Ghosting On Metal / Glass / And Ceramic Surfaces. Compact / Durable / IP52 Protection / Drop Resistance From 1.8m Ensures Reliable Operation In Industrial Environments. Ideal For Inventory Management / Electronics / Home Appliance / And Automotive Parts Handling.