Learn More
MSC AbilityOne Shop Towels, 12.5 x 13.4 in, White, 475 Sheets/Roll
Catalog No. 502818257
Encompass
These industrial shop towels are designed for general purpose cleaning and wiping tasks. Each roll contains 475 flat sheets, featuring a hydroentangled construction for durability.
  • General purpose wipe
  • Flat sheet form
  • Dry wipe style
  • Roll packaging type
  • Hydroentangled construction
  • White wipe color
  • Unscented
Catalog No. 50-281-8257 Supplier MSC Supplier No. 18995951
These industrial shop towels are designed for general purpose cleaning and wiping tasks. Each roll contains 475 flat sheets, featuring a hydroentangled construction for durability.
  • General purpose wipe
  • Flat sheet form
  • Dry wipe style
  • Roll packaging type
  • Hydroentangled construction
  • White wipe color
  • Unscented