Learn More
Lascar Electronics Inc USB Industrial Temperature Data Logger
SDP
Catalog No. NC1856436
Encompass_Preferred
This standalone data logger measures more than 32,000 readings over a -40 to +125°C (-40 to +257°­­F) range. Its robust stainless steel assembly provides protection from corrosion, impact and water ingress to IP67/NEMA 4X. Easily set up the logger and view downloaded data by plugging the unit into a PC’s USB port and using the free EasyLog software provided. Data can then be graphed, printed and exported to other applications for detailed analysis. Supplied with a specialist high temperature ⅔ AA battery.
Catalog No. NC1856436 Supplier Lascar Electronics Inc Supplier No. ELUSB1PRO
This standalone data logger measures more than 32,000 readings over a -40 to +125°C (-40 to +257°­­F) range. Its robust stainless steel assembly provides protection from corrosion, impact and water ingress to IP67/NEMA 4X. Easily set up the logger and view downloaded data by plugging the unit into a PC’s USB port and using the free EasyLog software provided. Data can then be graphed, printed and exported to other applications for detailed analysis. Supplied with a specialist high temperature ⅔ AA battery.