Learn More
Analytical West Analytical West, 10-60061, 1/4" OD x 0.007" Wall 316L Stainless Steel Tubing, 36" Length
SDP
Catalog No. NC1098001
Encompass_Preferred
The 10-60061 is a digital thermometer from Analytical West that measures temperatures from -50 to 150 degrees Celsius with 0.1 degree resolution. It has a large LCD display, max/min temperature recording, auto-off function, and a stainless steel penetration probe. The thermometer is powered by a single AAA battery and has an accuracy of ±0.5°C from -20 to 100°C. It comes with a soft carrying pouch and instructions. The 10-60061 is ideal for various temperature measurement applications including food, HVAC, industrial and more.
Catalog No. NC1098001 Supplier Analytical West Supplier No. 1060061
The 10-60061 is a digital thermometer from Analytical West that measures temperatures from -50 to 150 degrees Celsius with 0.1 degree resolution. It has a large LCD display, max/min temperature recording, auto-off function, and a stainless steel penetration probe. The thermometer is powered by a single AAA battery and has an accuracy of ±0.5°C from -20 to 100°C. It comes with a soft carrying pouch and instructions. The 10-60061 is ideal for various temperature measurement applications including food, HVAC, industrial and more.