Learn More
Description
- With gas inlet and outlet
- Not in conjunction with access port or interior lighting option
Specifications
Specifications
| Description | Inert gas connection - With gas inlet and outlet, Ø 10 mm, incl. additional measures for the greatest possible impermeability to gas; not in conjunction with access port or interior lighting option |
| For Use With (Equipment) | BINDER Chambers FD 56 |
| Material | Steel |
| Type | Gas Connection |
| Diameter (English) | 0.4 in. |
| Diameter (Metric) | 10 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
