Learn More
BINDER™ Inert Gas Connection with Gas Inlet and Outlet
SDP
Catalog No. 08101237 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Series FP 115 E3 Avantgarde
BINDER Series FP 260 Avantgarde
BINDER Series FP Avantgarde
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
08-101-237 BINDER Series FP 260 Avantgarde
08-101-235 BINDER Series FP Avantgarde
08-101-236 BINDER Series FP 115 E3 Avantgarde
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 08-101-237 Supplier BINDER™ Supplier No. 80122197
Add to Cart
Add to Cart
This item is not returnable. View return policy
The inert gas connection includes additional measures for the greatest possible impermeability to gas.
  • Accessory for Series FP Avantgarde Line Drying and Heating Chambers
Specifications
For Use With (Equipment) BINDER Series FP 260 Avantgarde
Type Connection Kit
Includes Gas Inlet and Outlet
Diameter (English) 0.39 in.
Diameter (Metric) 10 mm
Voltage 120/230 V