Learn More
Perkin Elmer US LLC Inert Gas Purifier - 0.125 in, Helium
Catalog No. 502103982
Encompass_Preferred
The mobile phase in gas chromatography transports analytes through the column, between the injector and detector, when the analytes are not partitioned into the stationary phase. This gas purifier removes hydrocarbons, moisture, and oxygen. Specifically for helium gas.
Catalog No. 50-210-3982 Supplier Perkin Elmer US LLC Supplier No. N6471025
The mobile phase in gas chromatography transports analytes through the column, between the injector and detector, when the analytes are not partitioned into the stationary phase. This gas purifier removes hydrocarbons, moisture, and oxygen. Specifically for helium gas.