Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Inert in-line inlet check valve; 0.014 Cv, PEEK/EPDM
Catalog No. NC1638705
Encompass
Unlike spring-actuated check valves that may restrict or impede the flow path, this in-line check valve features a smooth flow path that minimizes shear and turbulence.
Catalog No. NC1638705 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0135512
Unlike spring-actuated check valves that may restrict or impede the flow path, this in-line check valve features a smooth flow path that minimizes shear and turbulence.