Learn More
GSC Go Science Crazy Inertial Balance
SCI_ED
Catalog No. S13996 Shop All GSC Products
Click to view available options
Includes:
Instructions, 3 cylindrical masses, and 1 C-clamp
Catalog No. Includes
S13996 Instructions, 3 cylindrical masses, and 1 C-clamp
1 options
Catalog No. S13996 Supplier GSC Go Science Crazy Supplier No. INRTBL
Demonstrate Newton's First Law. The GSC Inertial Balance allows for the measurement of the inertial mass of an object.
  • Allows for the measurement of the inertial mass of an object, and is a effictive way to demonstrate Newton's First Law
  • The Inertial Balance allows for the comparison of both known and unknown masses
Specifications
Description Allows for the measurement of the inertial mass of an object
Includes Instructions, 3 cylindrical masses, and 1 C-clamp
Dimensions (L x W) 12.6 x 5.25 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov