Learn More
GL Sciences Inc Inertsil Hybrid-C18, 3.5 um 2.1 x 30 mm
Catalog No. 5000807357
Encompass
High performance C18 HPLC column based on hybrid silica technology designed for robust operation across a wide pH range. Suitable for both acidic and basic mobile phases providing excellent durability and reproducibility. Ideal for method transfer from commonly used columns such as BEH C18 CSH C18 XBridge and Gemini columns. Delivers consistent retention peak shape and long column lifetime for pharmaceutical environmental and general analytical applications.
Catalog No. 50-008-07357 Supplier GL Sciences Inc Supplier No. 502076026
High performance C18 HPLC column based on hybrid silica technology designed for robust operation across a wide pH range. Suitable for both acidic and basic mobile phases providing excellent durability and reproducibility. Ideal for method transfer from commonly used columns such as BEH C18 CSH C18 XBridge and Gemini columns. Delivers consistent retention peak shape and long column lifetime for pharmaceutical environmental and general analytical applications.