Learn More
GL Sciences Inc Inertsil ODS-3, 3 Micron, Conventional Guard Column, 10 X 4.0mm
Catalog No. NC1766574
Encompass
Virtually perfect peak shape for most bases and acids. Predictable selectivity with very low operating pressure making this ideal for fast/LC, UPLC, and LC/MS. Exceptional physical and chemical durability provides long column life. Vast database of chromatographic applications available to assist in method development.
Catalog No. NC1766574 Supplier GL Sciences Inc Supplier No. 502003655
Virtually perfect peak shape for most bases and acids. Predictable selectivity with very low operating pressure making this ideal for fast/LC, UPLC, and LC/MS. Exceptional physical and chemical durability provides long column life. Vast database of chromatographic applications available to assist in method development.