Learn More
GL Sciences Inc Inertsil ODS-3 Analytical Column, 5um, 4.6x150mm
Catalog No. NC1805725
Encompass
Inertsil ODS-3 is still GL Sciences’ most popular phase and continues to be used world widely and reliably for long established methods in pharmaceutical, and contract research labs. Inertsil ODS-3 has a relatively strong retentivity compared to commercially available ODS columns. In addition, the introduction of higher surface area silica provide high preparative loading capacity without sacrificing peak shape.
This item is not returnable. View return policy
Catalog No. NC1805725 Supplier GL Sciences Inc Supplier No. 502001731
Inertsil ODS-3 is still GL Sciences’ most popular phase and continues to be used world widely and reliably for long established methods in pharmaceutical, and contract research labs. Inertsil ODS-3 has a relatively strong retentivity compared to commercially available ODS columns. In addition, the introduction of higher surface area silica provide high preparative loading capacity without sacrificing peak shape.