Learn More
GL Sciences Inc Inertsil ODS-3V Analytical Column, 5um, 4.6x150mm
Catalog No. NC1730142
Encompass
Inertsil ODS-3V offers all of the outstanding chromatographic benefits of Inertsil ODS-3 with the added benefit of a more thoroughly documented, validated QC procedure consistent with the demands of GLP/GMP compliance. Each Inertsil ODS-3V is delivered with a Manufacturer‘s Validation Certificate showing the detailed results of every QA and QC step in manufacturing.
Catalog No. NC1730142 Supplier GL Sciences Inc Supplier No. 502001801
Inertsil ODS-3V offers all of the outstanding chromatographic benefits of Inertsil ODS-3 with the added benefit of a more thoroughly documented, validated QC procedure consistent with the demands of GLP/GMP compliance. Each Inertsil ODS-3V is delivered with a Manufacturer‘s Validation Certificate showing the detailed results of every QA and QC step in manufacturing.