Learn More
GL Sciences Inc Inertsil ODS-3V, 5 micron, 250 x 4.6mm
Catalog No. NC1733208
Encompass
Inertsil ODS-3V offers all of the outstanding chromatographic benefits of Inertsil ODS-3 with the added benefit of a more thoroughly documented, validated QC procedure consistent with the demands of GLP/GMP compliance. Each Inertsil ODS-3V is delivered with a Manufacturer's Validation Certificate showing the detailed results of every QA and QC step in manufacturing.The use of Inertsil ODS-3V columns provides an extra measure of assurance of consistent performance from column to column and batch to batch. Inertsil ODS-3V columns are also available in 3-column-sets packed with your choice of 3 different silica batches or a single silica batch to assist in reproducibility studies.
Catalog No. NC1733208 Supplier GL Sciences Inc Supplier No. 502001802
Inertsil ODS-3V offers all of the outstanding chromatographic benefits of Inertsil ODS-3 with the added benefit of a more thoroughly documented, validated QC procedure consistent with the demands of GLP/GMP compliance. Each Inertsil ODS-3V is delivered with a Manufacturer's Validation Certificate showing the detailed results of every QA and QC step in manufacturing.The use of Inertsil ODS-3V columns provides an extra measure of assurance of consistent performance from column to column and batch to batch. Inertsil ODS-3V columns are also available in 3-column-sets packed with your choice of 3 different silica batches or a single silica batch to assist in reproducibility studies.