Learn More
GL Sciences Inc Inertsil ODS-4, 3 Micron, 250 X 4.6mm
Catalog No. NC1791476
Encompass
Inertsil ODS-4 is highly inert ODS column as well as InertSustain C18. However, as the base Silica gel and carbon load are different, their selectivities show some differences according to the characteristics of the analytes. Inertsil ODS-4 was developed as a more inert alternative to Inertsil ODS-3 by applying upgraded deactivating techniques.
Catalog No. NC1791476 Supplier GL Sciences Inc Supplier No. 502004046
Inertsil ODS-4 is highly inert ODS column as well as InertSustain C18. However, as the base Silica gel and carbon load are different, their selectivities show some differences according to the characteristics of the analytes. Inertsil ODS-4 was developed as a more inert alternative to Inertsil ODS-3 by applying upgraded deactivating techniques.