Learn More
GL Sciences Inc Reversed Phase Columns (C18 Column), 5 um, 4.6 mm, 250 mm
Catalog No. NC1831616
Encompass
Inertsil ODS-4 delivers the same extreme inertness to any type of compounds just like InertSustain C18 along with unprecedented stability under 100 % aqueous mobile phases for qualitative and quantitative analysis. However, as the base silica gel and carbon loading are different on Inertsil ODS-4, differences in selectivity can be observed for certain analytes.
Catalog No. NC1831616 Supplier GL Sciences Inc Supplier No. 502003946
Inertsil ODS-4 delivers the same extreme inertness to any type of compounds just like InertSustain C18 along with unprecedented stability under 100 % aqueous mobile phases for qualitative and quantitative analysis. However, as the base silica gel and carbon loading are different on Inertsil ODS-4, differences in selectivity can be observed for certain analytes.