Learn More
GL Sciences Inc Inertsil ODS-2 Analytical Column, 5um, 4.6x250mm
Catalog No. NC1712791
Encompass
Inertsil ODS-2 columns have a pore size of 150 Å offering symmetric peaks for bases, acids with low pressure. When Inertsil ODS-2 was introduced in 1987s, this HPLC transformed the entire industry. Inertsil ODS-2 was the first HPLC phase created using ultra high purity silica, which produced superior base deactivation. Inertsil ODS-2 is one of the most popular phase and continues to be used widely and reliably for long established methods in pharmaceutical and environmental labs.
Catalog No. NC1712791 Supplier GL Sciences Inc Supplier No. 502001128
Inertsil ODS-2 columns have a pore size of 150 Å offering symmetric peaks for bases, acids with low pressure. When Inertsil ODS-2 was introduced in 1987s, this HPLC transformed the entire industry. Inertsil ODS-2 was the first HPLC phase created using ultra high purity silica, which produced superior base deactivation. Inertsil ODS-2 is one of the most popular phase and continues to be used widely and reliably for long established methods in pharmaceutical and environmental labs.