Learn More
GL Sciences Inc Inertsustain AQ-C18 3um
Catalog No. NC1669925
Encompass
Length 250, Diameter 4.6, Micron 3.
Catalog No. NC1669925 Supplier GL Sciences Inc Supplier No. 502089861
Length 250, Diameter 4.6, Micron 3.