Learn More
GL Sciences Inc Reversed Phase Columns (C18 Column), 3 um, 4.6 mm, 150 mm
Catalog No. NC1826107
Encompass
Generally, silica based columns are mechanically stable and provide high efficiencies, however, they cannot be used under alkaline conditions and their residual silanol groups tend to adsorb organic bases. InertSustain C18 employs a radically new type of silica, in which the surface of the silica is uniquely modified, enabling precise control of the silica properties.
Catalog No. NC1826107 Supplier GL Sciences Inc Supplier No. 502007445
Add to Cart
Add to Cart
Generally, silica based columns are mechanically stable and provide high efficiencies, however, they cannot be used under alkaline conditions and their residual silanol groups tend to adsorb organic bases. InertSustain C18 employs a radically new type of silica, in which the surface of the silica is uniquely modified, enabling precise control of the silica properties.