Learn More
Med Vet International Exel Butterfly Infusion Set, with 20g x 3/4in Needle, 12in Tubing, 50/Box
Catalog No. 502092289
Encompass_Preferred
Exel International’s butterfly infusion set, or scalp vein set has a 20g x 3/4in. thin wall stainless steel cannula, that is siliconized for easy insertion. It has a 12in. tube with flexible wings that hold the needle in position and helps to prevent inadvertent pull out. This infusion set has a yellow-colored hub for easy identification. Each scalp vein set in this box is latex free and individually packaged.
This item is not returnable. View return policy
Catalog No. 50-209-2289 Supplier Med Vet International Supplier No. 26703
Exel International’s butterfly infusion set, or scalp vein set has a 20g x 3/4in. thin wall stainless steel cannula, that is siliconized for easy insertion. It has a 12in. tube with flexible wings that hold the needle in position and helps to prevent inadvertent pull out. This infusion set has a yellow-colored hub for easy identification. Each scalp vein set in this box is latex free and individually packaged.