Learn More
Med Vet International Exel Butterfly Infusion Set, with 21g x 3/4in Needle, 12in Tubing, 50/Box
Catalog No. 502092290
Encompass_Preferred
Exel International’s butterfly infusion set, or scalp vein set, has a 21g x 3/4in. thin wall stainless steel cannula that is siliconized for easy insertion. It has a 12in. tube with flexible wings that hold the needle in position and helps prevent inadvertent pull out. This infusion set has a deep green-colored hub for easy identification. Each scalp vein set in this box is latex free and individually packaged.
This item is not returnable. View return policy
Catalog No. 50-209-2290 Supplier Med Vet International Supplier No. 26704
Exel International’s butterfly infusion set, or scalp vein set, has a 21g x 3/4in. thin wall stainless steel cannula that is siliconized for easy insertion. It has a 12in. tube with flexible wings that hold the needle in position and helps prevent inadvertent pull out. This infusion set has a deep green-colored hub for easy identification. Each scalp vein set in this box is latex free and individually packaged.