Learn More
Medchemexpress LLC HY-101868 5mg Medchemexpress, INF39 CAS:866028-26-4 Purity:>98%
SDP
Catalog No. 501967901
Encompass_Preferred
Medchemexpress, HY-101868 5mg INF39 CAS:866028-26-4 INF39 is an irreversible and noncytotoxic NLRP3 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7901 Supplier Medchemexpress LLC Supplier No. HY1018685MG
Medchemexpress, HY-101868 5mg INF39 CAS:866028-26-4 INF39 is an irreversible and noncytotoxic NLRP3 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.