Learn More
Azer Scientific Infant Heel Warmer
SDP
Catalog No. 22026735
Change view
Click to view available options
Quantity:
100
Catalog No. Quantity
22-026-735 100
1 options
This item is not returnable. View return policy
Catalog No. 22-026-735 Supplier Azer Scientific Supplier No. ES23532
This item is not returnable. View return policy
Azer Scientific Infant Heel Warmer is designed to safely and comfortably increase blood flow to an infant’s heel area, minimizing the potential for painful bruising, infection, or cartilage damage caused by frequent or repeated heel sticks.
  • Designed and tested for infant safety
  • Conforms to infant's foot for even heat distribution
  • Latex-free
  • Uniform heat distribution to improve blood flow to heel
  • Trigger disk allows for quick and easy activation
  • Available in liquid
  • Gently reach the optimal 104°F desired temperature.
Specifications
For Use With (Application) Infant needlesticks
Quantity 100