Learn More
Andwin Scientific INFECT/NON INFCT SHPR 50/CS
SDP
Catalog No. NC9077299
Encompass_Preferred
The STP-710 Disposable 2-Part Secondary Pressure Vessel is recommended for use with STP-110 & STP-310 Category A Shipping System. This product is sold in a case of 50.
Catalog No. NC9077299 Supplier Andwin Scientific Supplier No. STP710
The STP-710 Disposable 2-Part Secondary Pressure Vessel is recommended for use with STP-110 & STP-310 Category A Shipping System. This product is sold in a case of 50.