Learn More
Berlin Packaging INFECTIOUS SHIPPER KIT
Catalog No. NC1413452
Encompass_Preferred
Carton: 16 5/8” x 16 5/8” x 18 ½”275# Double Wide Kla White Corrugated4G/Class 6.2/YRCharcoalDry Ice Infectious ShipperCATEGORY A FOR 12X15 VIALSwithout Vials and Dry Ice
Catalog No. NC1413452 Supplier Berlin Packaging Supplier No. HMS69050
Carton: 16 5/8” x 16 5/8” x 18 ½”275# Double Wide Kla White Corrugated4G/Class 6.2/YRCharcoalDry Ice Infectious ShipperCATEGORY A FOR 12X15 VIALSwithout Vials and Dry Ice