Learn More
MSC Multi-Clean Floor Stripper, Water-Based, Lemon Scent, pH 11-12
Catalog No. 502838356
Encompass
This water-based floor finish stripper is designed for effective removal of various floor finishes. It features a lemon scent and a high pH of 11-12, making it suitable for a wide range of surfaces including concrete, terrazzo, vinyl, and vinyl composite tile.
  • Water-based formula for efficient floor finish removal
  • Clear liquid with a lemon scent
  • Complete solubility for easy application and rinse
  • High pH (11-12) for powerful stripping action
  • Suitable for use on concrete, terrazzo, vinyl, and vinyl composite tile (VCT)
Catalog No. 50-283-8356 Supplier MSC Supplier No. 73061848
This water-based floor finish stripper is designed for effective removal of various floor finishes. It features a lemon scent and a high pH of 11-12, making it suitable for a wide range of surfaces including concrete, terrazzo, vinyl, and vinyl composite tile.
  • Water-based formula for efficient floor finish removal
  • Clear liquid with a lemon scent
  • Complete solubility for easy application and rinse
  • High pH (11-12) for powerful stripping action
  • Suitable for use on concrete, terrazzo, vinyl, and vinyl composite tile (VCT)