Learn More
Medchemexpress LLC HY-13311 50mg Medchemexpress, Infigratinib CAS:872511-34-7 Purity:>98%
SDP
Catalog No. 502028980
Encompass_Preferred
Medchemexpress, HY-13311 50mg Infigratinib CAS:872511-34-7 Infigratinib (BGJ-398; NVP-BGJ398) is a potent inhibitor of the FGFR family with IC50s of 0.9 nM, 1.4 nM, 1 nM, and 60 nM for FGFR1, FGFR2, FGFR3, and FGFR4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8980 Supplier Medchemexpress LLC Supplier No. HY1331150MG
Medchemexpress, HY-13311 50mg Infigratinib CAS:872511-34-7 Infigratinib (BGJ-398; NVP-BGJ398) is a potent inhibitor of the FGFR family with IC50s of 0.9 nM, 1.4 nM, 1 nM, and 60 nM for FGFR1, FGFR2, FGFR3, and FGFR4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.