Learn More
Medchemexpress LLC Infigratinib-d3 | 98.9% | 563.49 | C26H28D3Cl2N7O3 | 5 MG
SDP
Catalog No. 5000262548
Encompass_Preferred
Infigratinib-d3 is the deuterated analog of the FGFR inhibitor infigratinib, supplied as a labeled standard for laboratory research. Provided in milligram quantities, it is intended for research use only and is commonly used as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS analyses.
  • Deuterated analog suitable as tracer or internal standard.
  • High chemical purity (98.9%).
  • Compatible with NMR, GC-MS, and LC-MS quantitative workflows.
  • Supplied in small mg-scale vials for analytical use.
  • Useful for calibration and quantitative assay development.
  • For research use only; not for human or animal therapeutic use.
Catalog No. 50-002-62548 Supplier Medchemexpress LLC Supplier No. HY13311S5MG
Infigratinib-d3 is the deuterated analog of the FGFR inhibitor infigratinib, supplied as a labeled standard for laboratory research. Provided in milligram quantities, it is intended for research use only and is commonly used as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS analyses.
  • Deuterated analog suitable as tracer or internal standard.
  • High chemical purity (98.9%).
  • Compatible with NMR, GC-MS, and LC-MS quantitative workflows.
  • Supplied in small mg-scale vials for analytical use.
  • Useful for calibration and quantitative assay development.
  • For research use only; not for human or animal therapeutic use.