Learn More
Medchemexpress LLC Infigratinib phosphate | 1310746-10-1 | 99.6% | C26H34Cl2N7O7P | 10MG
SDP
Catalog No. 5000258719
Encompass_Preferred
Infigratinib phosphate is the phosphate salt form of infigratinib, a selective fibroblast growth factor receptor (FGFR) inhibitor used as a research tool in oncology and cell signaling studies. It is supplied as high-purity solid or as a ready-to-use DMSO solution, suitable for biochemical, cellular, and in vivo research applications.
  • Selective FGFR inhibition for target validation and pathway studies.
  • High purity (99.6%) suitable for sensitive assays.
  • Available as solid or 10 mM solution in DMSO for flexible use.
  • Molecular weight and formula provided for dosing and formulation calculations.
  • Applicable to in vitro cellular assays and in vivo pharmacology studies.
  • Provided with accompanying safety and technical documentation.
Catalog No. 50-002-58719 Supplier Medchemexpress LLC Supplier No. HY13311A10MG
Infigratinib phosphate is the phosphate salt form of infigratinib, a selective fibroblast growth factor receptor (FGFR) inhibitor used as a research tool in oncology and cell signaling studies. It is supplied as high-purity solid or as a ready-to-use DMSO solution, suitable for biochemical, cellular, and in vivo research applications.
  • Selective FGFR inhibition for target validation and pathway studies.
  • High purity (99.6%) suitable for sensitive assays.
  • Available as solid or 10 mM solution in DMSO for flexible use.
  • Molecular weight and formula provided for dosing and formulation calculations.
  • Applicable to in vitro cellular assays and in vivo pharmacology studies.
  • Provided with accompanying safety and technical documentation.