Learn More
Selleck Chemical LLC Infigratinib Phosphate E4474-25MG
SDP
Supplier:  Selleck Chemical LLC E447425MG
Encompass_Preferred
Infigratinib Phosphate (NVP-BGJ398 phosphate) is the phosphate salt form of infigratinib an orally bioavailable pan-inhibitor of human fibroblast growth factor receptors (FGFRs) with IC50 of 0 9 nM 1 4 nM 1 nM and 60 nM for FGFR1 FGFR2 FGFR3 and FGFR4 respectively It also has potential antiangiogenic antineoplastic and antitumor activities
Catalog No. 50-000-39032