Learn More
Description
- Monochrome CMOS sensor with 1 in. optical format
- 1.7 megapixel camera with fast frame rates of 96 fps at full resolution (1608 x 1104)
- Direct USB3 interface for fast image delivery and simplified connectivity
- Compact form factor
- Support for locking industrial micro USB on the back for power and control
- INFINITY ANALYZE microscopy software included with purchase. No annual license fees. (Camera requires INFINITY ANALYZE v7.0.3 or higher)
- Software support for Windows and Mac, along with third-party plugins for MetaMorph and MicroManager
- Selectable 8 or 12-bit pixel data
- Recommended coupler: 1x
Specifications
Specifications
| Type | USB Microscope Camera |
| Autoclavable | Not Autoclavable |
| Sterility | Non-sterile |
| Voltage | 5 V |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

