Learn More
Electron Microscopy Sciences INFINITY2 1.4 Megapixel USB 2.0 Microscopy Camera, CCD, Monochrome
SDP
Catalog No. 5032305
Encompass
Equipped with a high-quality, Sony CCD sensor, INFINITY2 CCD USB 2.0 cameras offer excellent sensitivity, high dynamic range, and a 12-bit digital output. Features include binning, auto white balance, full exposure control, programmable gain, sub-windowing and region of interest.
Sensor: CCD
Type: Monochrome
Catalog No. 50-323-05 Supplier Electron Microscopy Sciences Supplier No. 95106
Equipped with a high-quality, Sony CCD sensor, INFINITY2 CCD USB 2.0 cameras offer excellent sensitivity, high dynamic range, and a 12-bit digital output. Features include binning, auto white balance, full exposure control, programmable gain, sub-windowing and region of interest.
Sensor: CCD
Type: Monochrome