Learn More
Electron Microscopy Sciences INFINITY2 2.0 Megapixel USB 2.0 Microscopy Camera CCD, Color
SDP
Catalog No. 5032306
Encompass
Equipped with a high-quality, Sony CCD sensor, INFINITY2 CCD USB 2.0 cameras offer excellent sensitivity, high dynamic range, and a 12-bit digital output. Features include binning, auto white balance, full exposure control, programmable gain, sub-windowing and region of interest.
Sensor: CCD
Type: Color
Catalog No. 50-323-06 Supplier Electron Microscopy Sciences Supplier No. 95107
Equipped with a high-quality, Sony CCD sensor, INFINITY2 CCD USB 2.0 cameras offer excellent sensitivity, high dynamic range, and a 12-bit digital output. Features include binning, auto white balance, full exposure control, programmable gain, sub-windowing and region of interest.
Sensor: CCD
Type: Color