Learn More
ACCU-SCOPE™ INFINITY 3-3UR Monochrome Camera
SDP
Catalog No. 01189222 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Voltage:
5 VDC
Catalog No. Voltage
01-189-222 5 VDC
1 options
Catalog No. 01-189-222 Supplier ACCU-SCOPE™ Supplier No. AU280MCCD
The INFINITY 3-3UR scientific digital camera features a Sony ICX674 CCD sensor, offers 53 fps at full 2.8-megapixel resolution via a high-speed USB 3.0 interface. Designed for use in scientific and industrial applications, providing unmatched light sensitivity needed for low light applications.
  • Industry leading Sony EXview HAD II sensor technology
  • Monochrome ICX674 CCD sensor with 2/3 in. optical format providing a resolution of 1936 x 1456 using 4.54 x 4.54 μm pixels
  • Fastest frame rate possible for the ICX674 sensor. Up to 53 fps at full resolution, 66 fps (1920 x 1088), 109 fps at VGA (640 x 480)
  • High-speed USB 3.0 interface for fastest image delivery and simplified connectivity. USB 2.0 supported.
  • GPI/O for control of peripherals and synchronization
  • Region of Interest (ROI) option to provide higher frame rates
  • Selectable 8 or 14-bit pixel data
  • Selectable tap readout, matching sensor performance to your application
  • Multiple frame rates supported, each optimized for lowest noise performance
  • Software compatible with Windows 10, 8, 7, XP, Vista, Mac OS X 10.7, 32 and 64-bit operating systems.
  • Includes TWAIN and Direct X/Direct Show support
Specifications
Type USB Microscope Camera
Autoclavable Not Autoclavable
Sterility Non-sterile
Voltage 5 VDC