Learn More
Electron Microscopy Sciences INFINITY3 -1M 1.4 Megapixel Cooled USB 2.0 Microscopy Camera, CCD, Monochrome
SDP
Catalog No. 5032314
Encompass
For low light fluorescence applications, the INFINITY cooled CCD USB 2.0 cameras offer cooling to 25 degree C below ambient. The Sony ICX285 EXview HAD sensor has very a high dynamic range, excellent sensitivity and a 12-bit digital output. Features include binning, auto white balance, full exposure control, programmable gain, sub-windowing and region of interest.
Sensor: Cooled CCD
Type: Monochrome
Catalog No. 50-323-14 Supplier Electron Microscopy Sciences Supplier No. 95111
Add to Cart
Add to Cart
Request a Quote
For low light fluorescence applications, the INFINITY cooled CCD USB 2.0 cameras offer cooling to 25 degree C below ambient. The Sony ICX285 EXview HAD sensor has very a high dynamic range, excellent sensitivity and a 12-bit digital output. Features include binning, auto white balance, full exposure control, programmable gain, sub-windowing and region of interest.
Sensor: Cooled CCD
Type: Monochrome